Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorouracil, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602777 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-777 5 g Amber Glass Bottle
18-602-776 1 g Amber Glass Bottle
18-602-778 25 g Amber Glass Bottle
18-602-779 100 g Amber Glass Bottle
18-602-780 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-777 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. FL1275GM
Add to Cart
Add to Cart

FL127, 51-21-8, C4H3FN2O2

Spectrum™ Chemical Fluorouracil, USP belongs to a class of medications known as anti-metabolites that treat pre-cancerous and cancerous skin growths. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51-21-8
Molecular Formula C4H3FN2O2
Molecular Weight (g/mol) 130.08
MDL Number MFCD00006018
InChI Key GHASVSINZRGABV-UHFFFAOYSA-N
IUPAC Name 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES FC1=CNC(=O)NC1=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 5 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.