Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluosilicic acid, 25 wt.% aqueous solution

Catalog No. AC410710025
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 16961-83-4 | F6H2Si | 144.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluosilicic acid
Color Colorless
CAS 16961-83-4, 7732-18-5
Density 1.2700g/mL
Packaging Plastic bottle
Specific Gravity 1.27
Quantity 2.5 kg
Molecular Formula F6H2Si
Physical Form Liquid
Linear Formula H2SiF6
MDL Number MFCD00036289
Synonym hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure
InChI Key OHORFAFFMDIQRR-UHFFFAOYSA-P
SMILES [H+].[H+].F[Si-2](F)(F)(F)(F)F
IUPAC Name hexafluorosilicon(2-);hydron
Molecular Weight (g/mol) 144.091
PubChem CID 21863527
Formula Weight 144.09
Percent Purity 19 to 20.6% (total fluorine)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.