Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Flupirtine maleate
GREENER_CHOICESDP

Catalog No. 286710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-671-0 10 mg
28-675-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-671-0 Supplier Tocris Bioscience™ Supplier No. 2867/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

KV7 channel activator; also GABAA modulator and indirect NMDA antagonist

Flupirtine maleate is a non-opioid analgesic with muscle relaxant properties. Activates KV7 potassium channels, indirectly antagonizes NMDA receptors and modulates GABAA receptors. Exhibits neuroprotective actions in a model of cerebral ischemia in mice and reduces apoptosis and necrosis induced by noxious stimuli.

Chemical Identifiers

CAS 75507-68-5
Molecular Formula C19H21FN4O6

Specifications

Quantity 10 mg
Formula Weight 420.4
Purity >98%
Molecular Weight (g/mol) 420.397
Chemical Name or Material Flupirtine maleate
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.