Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flurbiprofen, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602790
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
10 kg
Packaging:
Amber Glass Bottle

F1128, 5104-49-4, C15H13FO2

Spectrum™ Chemical Flurbiprofen, USP is a non-steroidal anti-inflammatory drug used to treat pain and inflammation. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5104-49-4
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00079303
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
IUPAC Name 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Melting Range 114°-117°C
Packaging Amber Glass Bottle
Quantity 100 g
UN Number UN2811
Percent Purity 99 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Flurbiprofen
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.