Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Flutax 1
GSA_VAGREENER_CHOICESDP

Catalog No. 22261 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-261 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-261 Supplier Tocris Bioscience™ Supplier No. 2226/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fluorescent taxol derivative; microtubule stain

Flutax 1 is a a fluorescent taxol derivative that binds to the taxol microtubule binding site with high affinity (Ka ∼ 107M-1). Can be used for direct imaging of the microtubule cytoskeleton. Absorption, fluorescence and fluorescence decay of Flutax 1 in solution are pH sensitive. Excitation maximum ∼ 495 nm; emission maximum ∼ 520 nm.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 191930-58-2
Molecular Formula C71H66N2O21

Specifications

Quantity 1 mg
Formula Weight 1283.3
Purity >98%
Molecular Weight (g/mol) 1283.302
Chemical Name or Material Flutax 1
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.