Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fluxapyroxad Monoclonal Antibody (1G5)
GREENER_CHOICE

Catalog No. PIMA549692
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549692 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549692 Supplier Invitrogen™ Supplier No. MA549692
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Fluxapyroxad is a broad-spectrum fungicide that belongs to the chemical class of succinate dehydrogenase inhibitors (SDHI). It is used to control various fungal diseases in crops such as cereals, fruits, vegetables, and ornamentals. Fluxapyroxad works by inhibiting the respiratory chain in fungi, leading to the disruption of energy production and ultimately causing fungal cell death. This active ingredient has a high level of efficacy against a wide range of fungal pathogens and is known for its long-lasting protection against disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluxapyroxad
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 1G5
Concentration 3.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of FPX hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.