Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FLVCR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570059
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a member of the major facilitator superfamily of transporter proteins. The encoded protein is a heme transporter that may play a critical role in erythropoiesis by protecting developing erythroid cells from heme toxicity. This gene may play a role in posterior column ataxia with retinitis pigmentosa and the hematological disorder Diamond-Blackfan syndrome.
Specifications
FLVCR1 | |
Polyclonal | |
Unconjugated | |
FLVCR1 | |
9630055N22Rik; AXPC1; eline leukemia virus subgroup C cellular receptor 1; feline leukemia virus subgroup C cellular receptor 1; feline leukemia virus subgroup C receptor; Feline leukemia virus subgroup C receptor-related protein 1; FLVCR; FLVCR heme transporter 1; FLVCR protein; FLVCR1; Major facilitator superfamily domain containing 7B; Mfsd7b; PCA; PCARP | |
Rabbit | |
Affinity chromatography | |
RUO | |
226844 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B2RXV4 | |
FLVCR1 | |
synthetic peptide directed towards the middle region of human FLVCR1 (aa 223-272). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction