Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FLVCR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558195
Description
Immunogen sequence: KADLRRQKAN KETLENKLQE EEEESNTSKV PTAVSEDHL Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.
BAPTA ((1,2-bis(2-Aminophenoxy)ethane-N,N,N',N'- tetraacetic acid) is a highly selective calcium chelator. BAPTA has spawned many of the most familiar fluorescent indicators for calcium and other ions. BAPTA and its analogs including BAPTA-AM have proven most critical in cell physiology research where they have been essential in defining the mechanisms underlying inter- or intracellular calcium homeostasis and calcium dependent exocytosis. BAPTA and its analogs may also find application in the study of diseases in which the regulation of excitatory neurotransmitter release and uptake has been disturbed.
Specifications
FLVCR2 | |
Polyclonal | |
Unconjugated | |
FLVCR2 | |
C14orf58; calcium-chelate transporter; CCT; EPV; feline leukemia virus subgroup C cellular receptor family member 2; feline leukemia virus subgroup C cellular receptor family, member 2; feline leukemia virus subgroup C receptor-related protein 2; FLVCR2; FLVCRL14q; major facilitator superfamily domain containing 7C; MFSD7C; PVHH; transporter | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55640 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPI3 | |
FLVCR2 | |
Recombinant protein corresponding to Human FLVCR2. Recombinant protein control fragment (Product #RP-96557). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction