Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLVCR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599060 Supplier Invitrogen™ Supplier No. PA599060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLVCR2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

BAPTA ((1,2-bis(2-Aminophenoxy)ethane-N,N,N',N'- tetraacetic acid) is a highly selective calcium chelator. BAPTA has spawned many of the most familiar fluorescent indicators for calcium and other ions. BAPTA and its analogs including BAPTA-AM have proven most critical in cell physiology research where they have been essential in defining the mechanisms underlying inter- or intracellular calcium homeostasis and calcium dependent exocytosis. BAPTA and its analogs may also find application in the study of diseases in which the regulation of excitatory neurotransmitter release and uptake has been disturbed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLVCR2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FLVCR2
Gene Accession No. Q91X85, Q9UPI3
Gene Alias C14orf58; calcium-chelate transporter; CCT; EPV; feline leukemia virus subgroup C cellular receptor family member 2; feline leukemia virus subgroup C cellular receptor family, member 2; feline leukemia virus subgroup C receptor-related protein 2; FLVCR2; FLVCRL14q; major facilitator superfamily domain containing 7C; MFSD7C; PVHH; transporter
Gene Symbols FLVCR2
Host Species Rabbit
Immunogen Recombinant Human Feline leukemia virus subgroup C receptor-related protein 2 protein (1-100AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217721, 55640
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.