Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLVCR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, ProteinTech
SDP

Catalog No. 502238524 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8524 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8524 Supplier Proteintech Group Inc Supplier No. 267041AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aliquoting is unnecessary for -20°C storage.

FLVCR2 Polyclonal antibody specifically detects FLVCR2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen FLVCR2
Applications Western Blot
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FLVCR2
Gene Accession No. P60815, Q91X85, Q9UPI3
Gene Alias C14orf58; calcium-chelate transporter; CCT; EPV; feline leukemia virus subgroup C cellular receptor family member 2; feline leukemia virus subgroup C cellular receptor family, member 2; feline leukemia virus subgroup C receptor-related protein 2; FLVCR2; FLVCRL14q; major facilitator superfamily domain containing 7C; MFSD7C; PVHH; transporter
Gene Symbols FLVCR2
Host Species Rabbit
Immunogen FLVCR2 Fusion Protein Ag24928
Purification Method Antigen affinity chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217721, 314323, 55640
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.