Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLYWCH2 Antibody (OTI3G3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272341PCP
Description
FLYWCH2 Monoclonal specifically detects FLYWCH2 in Human samples. It is validated for Flow Cytometry.Specifications
FLYWCH2 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
FLYWCH2 | |
Full length recombinant protein of human FLYWCH2 (NP_612448) produced in HEK293T cells. | |
0.1 mL | |
114984.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI3G3 | |
Flow Cytometry | |
FLYWCH family member 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction