Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLYWCH2 Monoclonal Antibody (OTI3G3)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525523

Catalog No. PIMA525523


Only null left
Add to Cart

Description

Description

FLYWCH2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

FLYWCH2 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FLYWCH2
Monoclonal
0.35 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96CP2
FLYWCH2
Full length human recombinant protein of FLYWCH2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3G3
Unconjugated
FLYWCH2
FLYWCH family member 2; FLYWCH2
Mouse
Affinity Chromatography
RUO
114984
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.