Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562214 Supplier Invitrogen™ Supplier No. PA562214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELHFQAATYL CLLRSIRKHV ALHQEFHGKG ERSVEESAGL VGLKLPHQPG GKGWEP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 96%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FMC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FMC1
Gene Accession No. Q96HJ9
Gene Alias ATP synthase assembly factor FMC1, mitochondrial; C7orf55; C7orf55-LUC7L2; C7orf55-LUC7L2 protein; C7orf55-LUC7L2 readthrough; FMC1; Formation of mitochondrial complex V assembly factor 1 homolog; HSPC268; Protein FMC1 homolog
Gene Symbols C7orf55-LUC7L2, FMC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FMC1. Recombinant protein control fragment (Product #RP-100548).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100996928, 154791
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.