Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMDV Polyprotein (3D polymerase) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986074
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6074 Supplier Bioss Supplier No. BS4524R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FMDV Polyprotein (3D polymerase) is involved in foot-and-mouth disease virus (FMDV) which is a picornavirus that causes an acute vesicular disease of cloven-hoofed animals. This virus continues to be threat to livestock worldwide with outbreaks causing severe economic losses.

Specifications

Antigen FMDV Polyprotein (3D polymerase)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from FMDVC 3D polymerase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.