Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FMDV Polyprotein (VPg2 protein) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS4523R

Description
FMDV Polyprotein (VPg2 protein) is involved in foot-and-mouth disease virus (FMDV) which is a picornavirus that causes an acute vesicular disease of cloven-hoofed animals. This virus continues to be threat to livestock worldwide with outbreaks causing severe economic losses.Specifications
FMDV Polyprotein (VPg2 protein) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
KLH conjugated synthetic peptide derived from FMDVC Polyprotein (VPg2 protein). | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction