Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FMN1/Formin 1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13185R

Description
Plays a role in the formation of adherens junction and the polymerization of linear actin cables.Specifications
FMN1/Formin 1 | |
Polyclonal | |
Unconjugated | |
Fmn1 | |
Fmn; Fmn1; formin (limb deformity); formin 1; Formin-1; Ld; Limb deformity protein; Limb deformity protein homolog; RGD1306349; RGD1310100 | |
Rabbit | |
Protein A | |
RUO | |
342184 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q68DA7 | |
Fmn1 | |
KLH conjugated synthetic peptide derived from human FMN1/Formin 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction