Learn More
Invitrogen™ FMN1 Monoclonal Antibody (4F4)

Description
Peptide Sequence: MENVDNSLDG SDVSEPAKPE AGLEVAQSIL SKFSMKSLFG FTSKLESVNP EEEDAVLKAF HSLDVNPTSQ QDDSSNGLDP QEAGSRVSPD LGNDEKIASV ETESEGSQR.
Specifications
Specifications
| Antigen | FMN1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4F4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Fmn1 |
| Gene Accession No. | Q68DA7 |
| Gene Alias | FMN; Fmn1; formin (limb deformity); formin 1; formin-1; Ld; limb deformity protein; Limb deformity protein homolog; RGD1306349; RGD1310100 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.