Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552149 Supplier Invitrogen™ Supplier No. PA552149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFGSFETAFK DTGLGREVLP PDCSSTEAGG DGIRRPPSGL EHQQTGLSES HQDPEKHPEA EKDEMEKPAK RTCKQKPVSK VVAKVQDLSS QVQRVVKTHS KGKETIAIRP AAHAEFVPKA DLLT Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 42%.

Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fmn1
Gene Accession No. Q68DA7
Gene Alias FMN; Fmn1; formin (limb deformity); formin 1; formin-1; Ld; limb deformity protein; Limb deformity protein homolog; RGD1306349; RGD1310100
Gene Symbols Fmn1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FMN1. Recombinant protein control fragment (Product #RP-88599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.