Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589020 Supplier Invitrogen™ Supplier No. PA589020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NIDMPKTEPK GADPESPRRE EMGCNADQES QSGPGVPQTQ GGEVKPKSPE TALEAFKALF IRPPRKGTTA DTSELEALKR KMRHEKESLR AVFERSNSKP ADGPSDSKSP DHSLTEQDDR TPGRLQAVWP PPKTKDTEEK VGLKYT; Positive Samples: 293T, A-549, U-87MG, LO2, Mouse testis; Cellular Location: Cell junction, Cell membrane, Cytoplasm, Cytoplasmic side, Nucleus, Peripheral membrane protein, adherens junction.

Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Fmn1
Gene Accession No. Q05860, Q68DA7
Gene Alias FMN; Fmn1; formin (limb deformity); formin 1; formin-1; Ld; limb deformity protein; Limb deformity protein homolog; RGD1306349; RGD1310100
Gene Symbols Fmn1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-495 of human FMN1 (NP_0010966541).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14260, 342184
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.