Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FMN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598482
Description
FMN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
Specifications
| FMN1 | |
| Polyclonal | |
| Unconjugated | |
| Fmn1 | |
| FMN; Fmn1; formin (limb deformity); formin 1; formin-1; Ld; limb deformity protein; Limb deformity protein homolog; RGD1306349; RGD1310100 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 342184 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.91 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q68DA7 | |
| Fmn1 | |
| Recombinant Human Formin-1 protein (354-487AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction