Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMN2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen FMN2
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:4000, Immunohistochemistry-Paraffin 1:300-1:500
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224615SS
SDP
View Documents
Novus Biologicals
NBP224615SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224615
SDP
View Documents
Novus Biologicals
NBP224615
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

FMN2 Polyclonal specifically detects FMN2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FMN2
Western Blot 1:100-1:4000, Immunohistochemistry-Paraffin 1:300-1:500
Polyclonal
Purified
RUO
Human
formin 2
FMN2
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human and 77% homologous in mouse and rat.
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Signal Transduction
0.1 ml PBS with 0.05% Sodium Azide
56776
A synthetic peptide corresponding to amino acids 657-688 of human FMN2 was used as immunogen, Gene Bank no NP_064450.3.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.