Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565632 Supplier Invitrogen™ Supplier No. PA565632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVASDSGGGVS PALAAKASGA PAAADGFQNV FTGRTLLEKL FSQQENGPPE EAEKFCSRII AMGLLLPFSD CFREPCNQNA QTNAASFDQD QLYTWAAVSQ PTHSLDYSEG Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.

FMN2, a novel formin homology protein of the cappuccino subfamily, is highly conserved between evolutionarily diverse vertebrates. FMN2 bears a high degree of similarity to formin-1 which is the founding member of formin homology proteins. FMN2 plays a crucial role in maintenance of the meiotic spindle. The protein is widely expressed in human fetal brain, adult whole brain, hypothalamus, retina, pancreatic islet and germinal-center B cells. Pathological role of the protein is identified in various tumors like parathyloid tumor, glioblastoma, retinoblastoma and chondrosarcoma. FMN2 mutations are often a cause of unexplained infertility in women. The gene encoding for human FMN2 is localized in the chromosomal region 1q43.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMN2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FMN2
Gene Accession No. Q9NZ56
Gene Alias AU024104; FMN2; formin 2; formin-2; RP11-90L13.1
Gene Symbols FMN2
Host Species Rabbit
Immunogen Recombinant Human FMN2. Recombinant protein control fragment (Product #RP-106300).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56776
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.