Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FMNL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | FMNL1 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FMNL1 Polyclonal specifically detects FMNL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FMNL1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
752 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KNAVLEHMEELQEQVALLTERLRDAENESMAKIAELEKQLSQARKELETLRERFSESTAMGASRRPPEPEKAPPAAPTRPSALELKVEELEEKGLIRILRGPGDAVSIEILPVAVATPSGGDAPTPGVPTGSPSPDLAPA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
C17orf1BMGC133052, C17orf1KW-13, CLL-associated antigen KW-13, FHOD4, FMNLMGC1894, formin-like, formin-like 1, formin-like protein 1, Leukocyte formin, MGC21878 | |
FMNL1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title