Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555814 Supplier Invitrogen™ Supplier No. PA555814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KNAVLEHMEE LQEQVALLTE RLRDAENESM AKIAELEKQL SQARKELETL RERFSESTAM GASRRPPEPE KAPPAAPTRP SALELKVEEL EEKGLIRILR GPGDAVSIEI LPVAVATPSG GDAPTPGVPT GSPSPDLAPA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

The Formin-like protein 1 (FRL1) gene encodes a formin-related protein which has been implicated in morphogenesis, cyokinesis, and cell polarity. Formins are a conserved class of proteins expressed in all eukaryotes and have one DAD (diaphanous autoregulatory domain), one FH2 (formin homology 2) domain and one GBD/FH3 (Rho GTPase-binding / formin homology 3) domain. FRL1 is located in the cytoplasm and is highly expressed in the spleen, lymph node and bone marrow cells. FRL1 possibly has a role in the control of cell motility, survival of macrophages and cytoskeletal organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMNL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FMNL1
Gene Accession No. O95466
Gene Alias 8030453N10Rik; AI553564; C17orf1; C17orf1B; CLL-associated antigen KW-13; FHOD4; Fmnl; Fmnl1; Fnrl; formin like 1; formin-like 1; formin-like protein 1; formin-related gene in leukocytes; formin-related protein; Frl; FRL1; Frls; KW-13; Leukocyte formin; lymphocyte specific formin related protein
Gene Symbols FMNL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FMNL1. Recombinant protein control fragment (Product #RP-88597).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.