Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FMNL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546742
Description
Peptide sequence: EKQLAQEAKK LDAKTPSQRN KWQQQELIAE LRRRQAKEHR PVYEGKDGTI.
FMNL3, also known as Formin-like protein 3, is a 1028 amino acid containing protein involved in immunity or defense. This developmental protein is implicated in polarity control, invasion, migration, or metastasis through regulation of the Rho-related signaling pathway. It belongs to the formin homology family and contains a FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain and has high sequence identity to the mouse Wbp3 protein. Two different isoforms have been described. Itis suspectedbe involved in gastric cancer.
Specifications
FMNL3 | |
Polyclonal | |
Unconjugated | |
FMNL3 | |
2700073B04Rik; FBP11; FHOD3; FMNL3; Formin homology 2 domain-containing protein 3; formin like 3; formin-like 3; formin-like 3 protein; formin-like protein 3; Frl2; KIAA2014; mKIAA2014; WBP3; WBP-3; WW domain binding protein 3; WW domain-binding protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
91010 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IVF7 | |
FMNL3 | |
Synthetic peptide directed towards the C-terminal of human FMNL3 (aa 944-993). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction