Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMNL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551720 Supplier Invitrogen™ Supplier No. PA551720
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLDVKELGRG MELIRRECSI HDNSVLRNFL STNEGKLDKL QRDAKTAEEA YNAVVRYFGE SPKTTPPSVF FPVFVRFIRS YKEAEQENEA RKKQEEVMRE KQLAQEAKKL DAKTPSQRNK WQQQE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

FMNL3, also known as Formin-like protein 3, is a 1028 amino acid containing protein involved in immunity or defense. This developmental protein is implicated in polarity control, invasion, migration, or metastasis through regulation of the Rho-related signaling pathway. It belongs to the formin homology family and contains a FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain and has high sequence identity to the mouse Wbp3 protein. Two different isoforms have been described. Itis suspectedbe involved in gastric cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMNL3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FMNL3
Gene Accession No. Q8IVF7
Gene Alias 2700073B04Rik; FBP11; FHOD3; FMNL3; Formin homology 2 domain-containing protein 3; formin like 3; formin-like 3; formin-like 3 protein; formin-like protein 3; Frl2; KIAA2014; mKIAA2014; WBP3; WBP-3; WW domain binding protein 3; WW domain-binding protein 3
Gene Symbols FMNL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FMNL3. Recombinant protein control fragment (Product #RP-102030).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91010
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.