Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FMO Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114348
Description
This antibody specifically recognizes FMO in Oryza sativa subspecies japonica (Rice).
FMOs are one subfamily of class B external flavoprotein monooxygenases (EC 1.14.13), which belong to the family of monooxygenase oxidoreductases, along with the other subfamilies Baeyer-Villiger monooxygenases and microbial N-hydroxylating monooxygenases. FMO's are found in fungi, yeast, plants, mammals, and bacteria. Plant FMO's play a role in defending against pathogens and catalyze specific steps in the biosynthesis of auxin, a plant hormone. Plant FMO's also play a role in the metabolism of glucosinolates. These non-xenobiotic roles of plant FMO's suggest that other FMO functions could be identified in non-plant organisms.
Specifications
FMO | |
Polyclonal | |
Unconjugated | |
LOC4334994 | |
LOC4334994; Os04g0128900; OSNPB_040128900; probable indole-3-pyruvate monooxygenase YUCCA5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4334994 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
0 | |
LOC4334994 | |
Synthetic peptide within rice Flavin-containing monooxygenase aa 1-50. | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction