Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521555
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521555 Supplier Invitrogen™ Supplier No. PA521555
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (83%), Rat (82%), Dog (88%), Pig (86%), Rabbit (85%), Rhesus Monkey (98%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Metabolic N-oxidation of the diet-derived amino-trimethylamine (TMA) is mediated by flavin-containing monooxygenase and is subject to an inherited FMO3 polymorphism in man resulting in a small subpopulation with reduced TMA N-oxidation capacity resulting in fish odor syndrome Trimethylaminuria. Three forms of the enzyme, FMO1 found in fetal liver, FMO2 found in adult liver, and FMO3 are encoded by genes clustered in the 1q23-q25 region. Flavin-containing monooxygenases are NADPH-dependent flavoenzymes that catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMO1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene FMO1
Gene Accession No. Q01740
Gene Alias dimethylaniline monooxygenase [N-oxide-forming] 1; Dimethylaniline oxidase 1; fetal hepatic flavin-containing monooxygenase 1; flavin containing dimethylaniline monoxygenase 1; flavin containing monooxygenase 1; Flavin-containing monooxygenase 1; Flavin-containing monooxygenase 1 (fetal liver); FMO 1; FMO 1A1; FMO form 1; Fmo1; Fmo-1; hepatic flavin-containing monooxygenase (EC 1.14.13.8); hepatic flavin-containing monooxygenase (FMO); hepatic flavin-containing monooxygenase 1; monooxygenase (FMO) (EC 1.14.13.8); RFMO1A
Gene Symbols FMO1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FMO1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.