Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109732
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109732 Supplier Invitrogen™ Supplier No. PA5109732
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFISTRHGTW VMSRISEDGY PWDSVFHTRF RSMLRNVLPR TAVKWMIEQQ MNRWFNHENY GLEPQNKYIM KEPVLNDDVP SRLLCGAIKV KSTVKELTET SAIFEDGTVE ENIDVIIFAT GYSFSFPFLE DSLVKVENNM VSLYKYIFPA HLDKSTLACI GLIQPLGSIF PTAELQARWV TRVFKGLCSL PSERTMMMDI IKRNEKRIDL FGESQSQTLQ TNYVDYLDEL ALEIGAKPDF CSLLFKDPKL AVRLYFGPCN S.

The flavin-containing monooxygenases are NADPH-dependent enzymes that catalyze the oxidation of many drugs and xenobiotics. In most mammals, there is a flavin-containing monooxygenase that catalyzes the N-oxidation of some primary alkylamines through an N-hydroxylamine intermediate. However, in humans, this enzyme is truncated and is probably rapidly degraded. The protein encoded by this gene represents the truncated form and apparently has no catalytic activity. A functional allele found in African Americans has been reported, but no sequence evidence has been deposited to support the finding. This gene is found in a cluster with the FMO1, FMO3, and FMO4 genes on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMO2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Fmo2
Gene Accession No. Q99518
Gene Alias 2310008D08Rik; 2310042I22Rik; AW107733; Dimethylaniline monooxygenase [N-oxide-forming] 2; dimethylaniline oxidase 2; flavin containing dimethylaniline monoxygenase 2; flavin containing monooxygenase 2; flavin containing monooxygenase 2 (non-functional); FMO 1B1; FMO 2; FMO, pulmonary; FMO1B1; FMO2; Pulmonary flavin-containing monooxygenase 2
Gene Symbols Fmo2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 210-470 of human FMO2 (NP_001451.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2327
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.