Learn More
Invitrogen™ FMO3 Monoclonal Antibody (OTI3H1)

Description
FMO3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | FMO3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3H1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Fmo3 |
| Gene Accession No. | P31513 |
| Gene Alias | AW111792; Dimethylaniline monooxygenase [N-oxide-forming] 3; dimethylaniline oxidase 3; dJ127D3.1; flavin containing dimethylaniline monoxygenase 3; flavin containing monooxygenase 3; flavin-containing monooxygenase 3; flavin-containing monooxygenase FMO3; FM03; FMO 1D1; FMO 3; FMO form 2; FMO II; FMO3; FMOII; hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; MGC34400; TMAU; Trimethylamine monooxygenase; Unknown (protein for MGC:128044) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.