Learn More
FMO3 Mouse anti-Human, Mouse, Clone: OTI1G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810419

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The mammalian flavin-containing monooxygenases (FMO; EC 1.14.13.8) represent a multigene family whose gene products are localized in the endoplasmic reticulum of many tissues. These enzymes catalyze the NADPH-dependent oxidative metabolism of many drugs, pesticides, and other foreign compounds. Their substrates are soft nucleophiles with an electron-rich center, typically a nitrogen, sulfur or phosphorus-containing functional group, as the site for oxidative attack by the enzyme.Specifications
FMO3 | |
Monoclonal | |
Unconjugated | |
FMO3 | |
FMOII, TMAU, dJ127D3.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
14262, 2328 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P31513, P97501 | |
FMO3 | |
Human recombit protein fragment corresponding to amino acids 1-291 of human FMO3 produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.