Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529807 Supplier Invitrogen™ Supplier No. PA529807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (80%), Rat (81%), Dog (84%), Rabbit (81%), Rhesus Monkey (93%), Chimpanzee (100%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The mammalian flavin-containing monooxygenases (FMO; EC 1.14.13.8) represent a multigene family whose gene products are localized in the endoplasmic reticulum of many tissues. These enzymes catalyze the NADPH-dependent oxidative metabolism of many drugs, pesticides, and other foreign compounds. Their substrates are soft nucleophiles with an electron-rich center, typically a nitrogen, sulfur or phosphorus-containing functional group, as the site for oxidative attack by the enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMO3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Fmo3
Gene Accession No. P31513, P97501
Gene Alias AW111792; Dimethylaniline monooxygenase [N-oxide-forming] 3; dimethylaniline oxidase 3; dJ127D3.1; flavin containing dimethylaniline monoxygenase 3; flavin containing monooxygenase 3; flavin-containing monooxygenase 3; flavin-containing monooxygenase FMO3; FM03; FMO 1D1; FMO 3; FMO form 2; FMO II; FMO3; FMOII; hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; MGC34400; TMAU; Trimethylamine monooxygenase; Unknown (protein for MGC:128044)
Gene Symbols Fmo3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FMO3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14262, 2328
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.