Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553084 Supplier Invitrogen™ Supplier No. PA553084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IYKSVFSNSS KEMMCFPDFP FPDDFPNFMH NSKIQEYIIA FAKEKNLLKY IQFKTFVSSV NKHPDFATTG QWDVTTERDG KKESAVFDAV MVCSGHHVYP NLPK Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.

The mammalian flavin-containing monooxygenases (FMO; EC 1.14.13.8) represent a multigene family whose gene products are localized in the endoplasmic reticulum of many tissues. These enzymes catalyze the NADPH-dependent oxidative metabolism of many drugs, pesticides, and other foreign compounds. Their substrates are soft nucleophiles with an electron-rich center, typically a nitrogen, sulfur or phosphorus-containing functional group, as the site for oxidative attack by the enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMO3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fmo3
Gene Accession No. P31513
Gene Alias AW111792; Dimethylaniline monooxygenase [N-oxide-forming] 3; dimethylaniline oxidase 3; dJ127D3.1; flavin containing dimethylaniline monoxygenase 3; flavin containing monooxygenase 3; flavin-containing monooxygenase 3; flavin-containing monooxygenase FMO3; FM03; FMO 1D1; FMO 3; FMO form 2; FMO II; FMO3; FMOII; hepatic flavin-containing monooxygenase 3; hepatic flavin-containing monooxygenase-3; MGC34400; TMAU; Trimethylamine monooxygenase; Unknown (protein for MGC:128044)
Gene Symbols Fmo3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FMO3. Recombinant protein control fragment (Product #RP-89950).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.