Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FMO4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579276
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, Mouse Liver Tissue, SMMC Cell, HEPA Cell, A431 Cell, MCF-7 Cell. IHC: Rat Liver Tissue.
FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
Specifications
FMO4 | |
Polyclonal | |
Unconjugated | |
Fmo4 | |
D1Ertd532e; Dimethylaniline monooxygenase [N-oxide-forming] 4; dimethylaniline oxidase 4; flavin containing dimethylaniline monoxygenase 4; flavin containing monooxygenase 4; flavin-containing monooxygenase 4; flavin-containing monooxygenase FMO4; FMO 1E1; FMO 4; FMO2; Fmo4; Hepatic flavin-containing monooxygenase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
226564, 2329, 246247 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P31512, Q8K4B7, Q8VHG0 | |
Fmo4 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human Flavin containing monooxygenase 4 (75-92aa HEDYPNFMNHEKFWDYLQ). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction