Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMO4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989801
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9801 Supplier Bioss Supplier No. BS16099R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.

Specifications

Antigen FMO4
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene Fmo4
Gene Accession No. P31512
Gene Alias BOS_15742; D1Ertd532e; Dimethylaniline monooxygenase [N-oxide-forming] 4; Dimethylaniline oxidase 4; flavin containing monooxygenase 4; flavin-containing monooxygenase 4; flavin-containing monooxygenase FMO4; FMO 1E1; FMO 4; FMO2; Fmo4; Hepatic flavin-containing monooxygenase 4
Gene Symbols Fmo4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FMO4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2329
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.