Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMO5 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen FMO5
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159731
SDP
View Documents
Novus Biologicals
NBP159731
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

FMO5 Polyclonal specifically detects FMO5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FMO5
Unconjugated
RUO
P49326
2330
Synthetic peptides corresponding to FMO5(flavin containing monooxygenase 5) The peptide sequence was selected from the middle region of FMO5. Peptide sequence NKYLEKKINQRFDHEMFGLKPKHRALSQHPTLNDDLPNRIISGLVKVKGN.
Primary
Polyclonal
Rabbit
Cancer
dimethylaniline monooxygenase [N-oxide-forming] 5, Dimethylaniline oxidase 5, EC 1.14.13.8, flavin containing monooxygenase 5, FMO 5, Hepatic flavin-containing monooxygenase 5
FMO5
IgG
59 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.