Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Asp O-2-PhiPr -OH, Advanced ChemTech

Catalog No. 01340547
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 200336-86-3
Molecular Formula C28H27NO6
Molecular Weight (g/mol) 473.53
InChI Key MEAHCBOPNIDLFH-DEOSSOPVSA-N
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxo-4-[(2-phenylpropan-2-yl)oxy]butanoic acid
SMILES CC(C)(OC(=O)C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)C1=CC=CC=C1

Specifications

Color White to Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Fmoc-Asp O-2-PhiPr -OH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.