Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Glu OtBu -Thr psiMe.Mepro, Advanced ChemTech

Catalog No. 01339394
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-339-394

Supplier: Advanced Chem Tech PTE0191G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 957780-56-2
Molecular Formula C31H38N2O8
Molecular Weight (g/mol) 566.65
MDL Number MFCD18427360
InChI Key SIDKUUFAHNZJLW-VAQLEPBLSA-N
IUPAC Name (4S,5R)-3-[(2S)-5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
SMILES C[C@H]1OC(C)(C)N([C@@H]1C(O)=O)C(=O)[C@H](CCC(=O)OC(C)(C)C)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White to Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-Glu OtBu -Thr psiMe.Mepro
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.