Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMOC-Glycine, 98%, Thermo Scientific™

Catalog No. AC265230010
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC265230010 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC265230010 Supplier Thermo Scientific Chemicals Supplier No. 265230010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 174°C to 178°C
CAS 29022-11-5
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C17H15NO4
Linear Formula FMOC-NHCH2COOH
MDL Number MFCD00037140
Synonym fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine
InChI Key NDKDFTQNXLHCGO-UHFFFAOYSA-N
SMILES OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid
Quantity 1 g
Molecular Weight (g/mol) 297.31
PubChem CID 93124
Formula Weight 297.32
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material FMOC-Glycine, 98%
Show More Show Less
EINECSNumber 249-373-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.