Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMOC-L-Norleucine, 98%

Catalog No. AC442720010
Click to view available options
Quantity:
1 g

CAS: 77284-32-3 | C21H23NO4 | 353.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77284-32-3
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037537
InChI Key VCFCFPNRQDANPN-IBGZPJMESA-N
Synonym fmoc-nle-oh, fmoc-l-norleucine, n-fmoc-l-norleucine, s-2-fmoc-amino caproic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-norleucine, fmocnleoh, fmoc-nle
PubChem CID 7009636
SMILES CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 138°C to 142°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -19
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
Formula Weight 353.42
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material FMOC-L-Norleucine
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.