Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMOC-O-tert-Butyl-L-serine, 98%, Thermo Scientific™

Catalog No. ac265320010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 71989-33-8
Molecular Formula C22H24NO5
Molecular Weight (g/mol) 382.44
MDL Number MFCD00037127
InChI Key REITVGIIZHFVGU-IBGZPJMESA-M
Synonym fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh
PubChem CID 2724633
IUPAC Name (2S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
SMILES CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Melting Point 127°C to 131°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Specific Rotation Condition + 25.00 (24.00°C c=1,EtOAc)
Specific Rotation +24° to +26° (24°C, 589nm) (c=1, EtOAc)
Formula Weight 383.45
Percent Purity ≥97.5%
Physical Form Amorphous Powder
Chemical Name or Material FMOC-O-tert-Butyl-L-serine, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.