Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Thyronine, Advanced ChemTech

Catalog No. 01339876
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-339-876

Supplier: Advanced Chem Tech FX25201G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1354485-04-3
Molecular Formula C30H25NO6
Molecular Weight (g/mol) 495.53
InChI Key LFDWCUHMDBIXPE-NDEPHWFRSA-N
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[4-(4-hydroxyphenoxy)phenyl]propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(OC2=CC=C(O)C=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White to Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-Thyronine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.