Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMR1 Mouse anti-Mouse,Human,Rat, Unconjugated, Clone: 1F1, MilliporeSigma™

Catalog No. MABN24532MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN24532MI 25 μg
MABN24531MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN24532MI Supplier MilliporeSigma Supplier No. MABN245325UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FMR1 Monoclonal antibody specifically detects FMR1 in Mouse,Human,Rat samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMR1
Applications Western Blot
Classification Monoclonal
Clone 1F1
Conjugate Unconjugated
Gene Alias Synaptic functional regulator FMR1;Fragile X mental retardation protein 1 homolog;FMRP;Protein FMR-1;mFmr1p
Gene Symbols Fmr1, Fmr-1
Host Species Mouse
Immunogen Full-length recombinant mouse Fragile X mental retardation protein 1 homolog (FMR1).
Molecular Weight of Antigen ∼72 kDa observed; 68.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 1F1 is a mouse monoclonal antibody that specifically detects Fragile X mental retardation protein 1 homolog (FMR1). It targets an epitope with in the C-terminal half.
Target Species Mouse, Human, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.