Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FMR1NB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103507 Supplier Invitrogen™ Supplier No. PA5103507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FMR1NB.

FMR1NB gene ontology (GO) annotation includes integral component of membrane and integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMR1NB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FMR1NB
Gene Accession No. Q80ZA7, Q8N0W7
Gene Alias 3830422N12Rik; cancer/testis antigen 37; CT37; FMR1NB; fragile X mental retardation 1 neighbor; fragile X mental retardation 1 neighbor protein; NYSAR35; NY-SAR-35; Protein mNY-SAR-35; sarcoma antigen NY-SAR-35
Gene Symbols FMR1NB
Host Species Rabbit
Immunogen A synthesized peptide derived from human FMR1NB(Accession Q8N0W7), corresponding to amino acid residues F208-E255.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158521, 207854
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.