Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMR1polyG Mouse anti-Human, Clone: 8FM-2F7, MilliporeSigma™

Catalog No. MABN228025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2280-25 25 μL
MABN2280-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2280-25 Supplier MilliporeSigma Supplier No. MABN228025UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FMR1polyG, clone 8FM-2F7 is a highly specific mouse monoclonal antibody that targets Synaptic functional regulator FMR1 (FMR1polyG) and has been tested for use Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMR1polyG
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8FM-2F7
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols FMR1
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide with EAPLPGGVRQRGG sequence. Note: This protein is translated within the 5'UTR of the FMR1 gene and in a different frame resulting in a protein with different sequence and containing polyGlycine.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_002015
Test Specificity Clone 8FM-2F7 specifically detects FMR1polyG in human cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.