Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMRP Antibody (OTI1C6), PE/Atto594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270756PEATT594

Catalog No. N270756P594


Only null left
Add to Cart

Description

Description

FMRP Monoclonal specifically detects FMRP in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

FMRP
Monoclonal
PE/ATTO 594
PBS with 0.05% Sodium Azide
FMR1
Recombinant protein fragment corresponding to amino acids 36-279 of human FMRP (NP_002015) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat, Canine, Monkey
IgG1
Flow Cytometry
OTI1C6
Flow Cytometry
FMRPFRAXAPOF, fragile X mental retardation 1, fragile X mental retardation 1 protein, MGC87458, POF1, premature ovarian failure 1, Protein FMR-1
Mouse
Affinity Purified
Epigenetics, microRNA
2332.0
Store at 4°C in the dark. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.