Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMRP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115243 Supplier Thermo Scientific Supplier No. PA5115243
Add to Cart
Add to Cart

FMRP Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen FMRP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FMR1
Gene Accession No. P35922, Q06787, Q80WE1
Gene Alias AT24755; BcDNA:GM08679; cg 6203 gene product from transcript cg6203-rc; cg6203; CG6203-PA; CG6203-PB; CG6203-PC; CG6203-PD; CG6203-PE; CG6203-PF; CG6203-PG; CG6203-PH; CG6203-PI; CG6203-PJ; CG6203-PK; dFMR; DFmr1; dFmrp; dfxr; dfxr1; dFXRP; Dmel\CG6203; Dmel_CG6203; dmfr1; drosophila fragile X mental retardation protein; EP(3)3517; FMR; FMR1; Fmr-1; Fmr1-PA; Fmr1-PB; Fmr1-PC; Fmr1-PD; Fmr1-PE; Fmr1-PF; Fmr1-PG; Fmr1-PH; Fmr1-PI; Fmr1-PJ; Fmr1-PK; FMRP; FMRP translational regulator 1; fragile X; fragile X mental retardation; fragile X mental retardation 1; fragile X mental retardation gene; fragile X mental retardation protein; fragile X mental retardation protein 1; Fragile X mental retardation protein 1 homolog; fragile X mental retardation related 1; fragile X mental retardation syndrome 1; fragile X mental retardation syndrome 1 homolog; Fragile X mental retardation syndrome-related protein 1; fragile X mental retardation-1 protein; fragile X protein; fragile x related; fragile X related protein; fragile X retardation 1 protein; fragile X-related; fragile-X; Fragile-X mental retardation 1; Fragile-X mental retardation protein; Fragile-X-related; FRAXA; FXR; MGC87458; POF; POF1; protein FMR-1; ragile X mental retardation protein; synaptic functional regulator FMR1
Gene Symbols FMR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FMR1(Accession Q06787), corresponding to amino acid residues Y476-L526.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14265, 2332, 24948
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.