Learn More
Promega FMS Kinase Enzyme System, 10microg
Catalog No. PRV4022 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4022 10 μg
1 options
Catalog No. PRV4022 Supplier Promega Supplier No. V4022
Recombinant human FMS (539-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FMS is a proto-oncogene that encodes the tyrosine kinase transmembrane receptor for colony stimulating factor 1 receptor (CSF1R).
  • Easily Screen and Profile FMS Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C