Learn More
Promega FMS Kinase Enzyme System, 1mg
Catalog No. PRV6395 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6395 1 mg
1 options
Catalog No. PRV6395 Supplier Promega Supplier No. V6395
Add to Cart
Add to Cart
Request a Quote
Recombinant human FMS (539-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FMS is a proto-oncogene that encodes the tyrosine kinase transmembrane receptor for colony stimulating factor 1 receptor (CSF1R).
  • Easily Screen and Profile FMS Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C