Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FN1 Mouse anti-Human, Clone: 568, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14255
Description
Mouse monoclonal antibody raised against native human FN1.
- High specificity
- High reactivity
Specifications
FN1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
P02751 | |
FN1 | |
High molecular weight proteins secreted by cultivated human fibroblasts. | |
100 μg | |
Primary | |
Epitope of this monoclonal antibody is located in the 8th type III repeat in the cell-binding region of fibronectin. | |
Store at 4°C. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
568 | |
Mouse monoclonal antibody raised against native human FN1. | |
In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
CIG/DKFZp686F10164/DKFZp686H0342/DKFZp686I1370/DKFZp686O13149/ED-B/FINC/FN/FNZ/GFND/GFND2/LETS/MSF | |
Mouse | |
Protein A/G purification | |
RUO | |
2335 | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction